Merck
  • TLR ligands upregulate RIG-I expression in human plasmacytoid dendritic cells in a type I IFN-independent manner.
TLR ligands upregulate RIG-I expression in human plasmacytoid dendritic cells in a type I IFN-independent manner.
Immunology and cell biology (2014-05-21)
Attila Szabo, Zoltan Magyarics, Kitti Pazmandi, Laszlo Gopcsa, Eva Rajnavolgyi, Attila Bacsi
ABSTRACT
Plasmacytoid dendritic cells (pDCs) are professional type I interferon (IFN)-producing cells that play an essential role in antiviral immunity. In many cell types, detection of intracellular pathogens is mostly dependent on endosomal Toll-like receptors (TLRs) and cytosolic sensors, such as retinoic acid-inducible gene I (RIG-I). However, the possible interplay between these two systems has not yet been elucidated. Here we aimed to study the collaboration of endosomal TLRs and RIG-I in primary human pDCs. We found that under steady-state conditions, pDCs express RIG-I at very low level, but the expression of this receptor is rapidly and dramatically upregulated upon stimulation by the TLR7 ligand imiquimod or the TLR9 ligand type A CpG. We also demonstrated that pDCs are able to sense and respond to 5'-triphosphate double-stranded RNA (5'-ppp-dsRNA) only following activation by endosomal TLRs. Experiments on primary pDCs with functionally blocked IFN-α/β receptor 1 (IFNAR1) and those on human pDC leukemia (pDC-L) cells defective in type I IFN secretion indicated that the upregulation of RIG-I expression in pDCs upon stimulation by endosomal TLR occurs in a type I IFN-independent manner. Selective phosphorylation of signal transducer and activator of transcription 1 (STAT1) on tyrosine 701 could be identified as an early signaling event in this process. Our results show that in contrast to many other cell types, where RIG-I expression is induced by type I IFN, in pDCs a disparate mechanism is responsible for the upregulation of RIG-I. Our findings also indicate that along with autophagy, an additional mechanism is operating in pDCs to promote the detection of replicating viruses.
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
IL-3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
IL-3 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Imiquimod, ≥98% (HPLC), solid