Merck
  • In vitro and in vivo evaluation of blood coagulation activation of polyvinyl alcohol hydrogel plus dextran-based vascular grafts.
In vitro and in vivo evaluation of blood coagulation activation of polyvinyl alcohol hydrogel plus dextran-based vascular grafts.
Journal of biomedical materials research. Part A (2014-07-22)
Nuno Alexandre, Elísio Costa, Susana Coimbra, Alice Silva, Ascensão Lopes, Miguel Rodrigues, Marta Santos, Ana Colette Maurício, José Domingos Santos, Ana Lúcia Luís
ABSTRACT
Polyvinyl alcohol hydrogel (PVA) is a water-soluble synthetic polymer that is commonly used in biomedical applications including vascular grafting. It was argued that the copolymerization of PVA with dextran (Dx) can result in improvement of blood-biomaterial interactions. The focus of this experimental study was to assess that interaction through an in vivo and in vitro evaluation of the coagulation system activation. The thrombogenicity of the copolymer was determined by quantification of platelet adhesion through the lactate dehydrogenase assay, determination of whole blood clotting time, and by quantification of platelet activation by flow cytometry. The thrombin-antithrombin complex blood levels were also determined. The obtained results for the in vitro assays suggested a non-thrombogenic profile for PVA/Dx. Additionally in vivo coagulation and hematological parameters were determined in an animal model after PVA/Dx vascular graft implantation. For coagulation homeostasis assessment, the intrinsic and extrinsic pathway's activation was determined by measuring prothrombin time (PT) and activated partial thromboplastin time (aPTT). Other markers of coagulation and inflammation activation including d-dimers, interleukin-6, and C-reactive protein were also assessed. The PVA/Dx copolymer tended to inhibit platelet adhesion/activation process and the contact activation process for coagulation. These results were also confirmed with the in vivo experiments where the measurements for APTT, interleukin-6, and C-reactive protein parameters were normal considering the species normal range of values. The response to those events is an indicator of the in vitro and in vivo hemocompatibility of PVA/Dx and it allows us to select this biomaterial for further preclinical trials in vascular reconstruction.
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, 1 M in H2O
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material