Merck
  • Super-stretchable graphene oxide macroscopic fibers with outstanding knotability fabricated by dry film scrolling.
Super-stretchable graphene oxide macroscopic fibers with outstanding knotability fabricated by dry film scrolling.
ACS nano (2014-05-07)
Rodolfo Cruz-Silva, Aaron Morelos-Gomez, Hyung-Ick Kim, Hong-Kyu Jang, Ferdinando Tristan, Sofia Vega-Diaz, Lakshmy P Rajukumar, Ana Laura Elías, Nestor Perea-Lopez, Jonghwan Suhr, Morinobu Endo, Mauricio Terrones
ABSTRACT
Graphene oxide (GO) has recently become an attractive building block for fabricating graphene-based functional materials. GO films and fibers have been prepared mainly by vacuum filtration and wet spinning. These materials exhibit relatively high Young's moduli but low toughness and a high tendency to tear or break. Here, we report an alternative method, using bar coating and drying of water/GO dispersions, for preparing large-area GO thin films (e.g., 800-1200 cm(2) or larger) with an outstanding mechanical behavior and excellent tear resistance. These dried films were subsequently scrolled to prepare GO fibers with extremely large elongation to fracture (up to 76%), high toughness (up to 17 J/m(3)), and attractive macroscopic properties, such as uniform circular cross section, smooth surface, and great knotability. This method is simple, and after thermal reduction of the GO material, it can render highly electrically conducting graphene-based fibers with values up to 416 S/cm at room temperature. In this context, GO fibers annealed at 2000 °C were also successfully used as electron field emitters operating at low turn on voltages of ca. 0.48 V/μm and high current densities (5.3 A/cm(2)). Robust GO fibers and large-area films with fascinating architectures and outstanding mechanical and electrical properties were prepared with bar coating followed by dry film scrolling.
Sigma-Aldrich
Sulfuric acid, 99.999%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)