Merck
  • Cholinergic receptor blockade by scopolamine and mecamylamine exacerbates global cerebral ischemia induced memory dysfunction in C57BL/6J mice.
Cholinergic receptor blockade by scopolamine and mecamylamine exacerbates global cerebral ischemia induced memory dysfunction in C57BL/6J mice.
Nitric oxide : biology and chemistry (2014-08-30)
R S Ray, S Rai, A Katyal
ABSTRACT
Global cerebral ischemia/reperfusion (GCI/R) injury encompasses complex pathophysiological sequalae, inducing loss of hippocampal neurons and behavioural deficits. Progressive neuronal death and memory dysfunctions culminate from several different mechanisms like oxidative stress, excitotoxicity, neuroinflammation and cholinergic hypofunction. Experimental evidences point to the beneficial effects of cholinomimetic agents such as rivastigmine and galantamine in improving memory outcomes following GCI/R injury. However, the direct implications of muscarinic and nicotinic receptor blockade during global cerebral ischemia/reperfusion injury have not been investigated. Therefore, we evaluated the relative involvement of muscarinic and nicotinic receptors in spatial/associative memory functions and neuronal damage during global cerebral ischemia reperfusion injury. The outcomes of present study support the idea that preservation of both muscarinic and nicotinic receptor functions is essential to alleviate hippocampal neuronal death in CA1 region following global cerebral ischemia/reperfusion injury.
Product Description
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Supelco
Mycophenolic acid, analytical standard
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)