Merck
  • Effect of component distribution and nanoporosity in CuPt nanotubes on electrocatalysis of the oxygen reduction reaction.
Effect of component distribution and nanoporosity in CuPt nanotubes on electrocatalysis of the oxygen reduction reaction.
ChemSusChem (2014-12-17)
Huizhang Guo, Xiang Liu, Chengdong Bai, Yuanzhi Chen, Laisen Wang, Mingsen Zheng, Quanfeng Dong, Dong-Liang Peng
ABSTRACT
Pt-based bimetallic electrocatalysts hold great potential in the oxygen reduction reaction (ORR) in current fuel-cell prototypes. However, they also face challenges from drastic dealloying of less-noble metals and coalescence of small nanoparticles. Porous and structure-ordered nanotubes may hold the potential to improve the stability of bimetallic electrocatalysts. Herein, we report a method to prepare CuPt nanotubes and porous Cu3 Pt intermetallic nanorods through a controlled galvanic replacement reaction and heat treatment process. The effect of the geometric features and compositional segregation on the electrocatalysis of the ORR was clarified. The outstanding performance of the Cu3 Pt/C-700 catalyst in the ORR relative to that of CuPt/C-RT was mainly attributed to the nanoporosity of the catalyst, whereas the enhanced specific activity on CuPt/C-RT after potential cycling was attributed to the interaction between the CuPt alloyed core and the Pt shell in the tube wall.
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
Nickel(II) acetylacetonate, 95%
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Supelco
Toluene, analytical standard
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%