Merck
  • Autocrine VEGF/VEGFR1 signaling in a subpopulation of cells associates with aggressive osteosarcoma.
Autocrine VEGF/VEGFR1 signaling in a subpopulation of cells associates with aggressive osteosarcoma.
Molecular cancer research : MCR (2014-04-25)
Tetsuro Ohba, Justin M M Cates, Heather A Cole, David A Slosky, Hirotaka Haro, Takashi Ando, Herbert S Schwartz, Jonathan G Schoenecker
ABSTRACT
Osteosarcoma is the most common primary bone malignancy and accounts for more than half of primary skeletal malignancies in children and young adults. Although vascular endothelial growth factor (VEGF) expression in osteosarcoma has been associated with poor outcome, its role in the pathogenesis of osteosarcoma remains controversial. Here, VEGF and VEGFR1 expression in both human and murine osteosarcoma cells associated with increasing malignant potential. Autocrine VEGF/VEGFR1 signaling resulted in constitutive activation of VEGFR1 in highly aggressive osteosarcoma cells. In addition, survival and proliferation of highly aggressive osteosarcoma cells was dependent on autocrine VEGF/R1 signaling in vitro. The effect of VEGFR1 expression on in vivo tumor growth and angiogenesis was evaluated by immunoselecting subpopulations of osteosarcoma cells that express high or low levels of VEGFR1. Cell enriched for high VEGFR1 expression showed increased VEGF production, tumor growth, tumor angiogenesis, and osteolysis in vivo. In addition, it was demonstrated that VEGF and VEGFR1 are coexpressed by a subset of tumor cells in human osteosarcoma, similar to what was observed in the murine osteosarcoma cells. These results suggest that autocrine VEGF/VEGFR1 signaling in a subpopulation of tumor cells plays a pivotal role in osteosarcoma progression. Aggressive osteosarcoma phenotypes are mediated by autocrine VEGF/VEGFR1 signaling and improved stratification measures and novel anti-angiogenic strategies may benefit this specific tumor type.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material