Merck
  • IL-21 receptor signalling partially mediates Th2-mediated allergic airway responses.
IL-21 receptor signalling partially mediates Th2-mediated allergic airway responses.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2014-05-09)
S Lajoie, I Lewkowich, N S Herman, A Sproles, J T Pesce, T A Wynn, M J Grusby, Q Hamid, M Wills-Karp
ABSTRACT
Interleukin-21 (IL-21) has been implicated in the development of Th2-mediated immune responses; however, the exact role it plays in allergic diseases is not well understood. To elucidate the contribution of IL-21 receptor signalling to Th2-dependent immune responses in the lung. We compared allergic airway responses in wild-type BALB/c and Il21r-deficient mice exposed to local airway challenge with house dust mite (HDM). We demonstrate that IL-21R-deficiency reduces HDM-driven airway hyperresponsiveness (AHR) with only partial effects on airway inflammation. Concomitant with the reduction in AHR in Il21r-deficient mice, significant suppression was observed in protein levels of the Th2 cytokines IL-4, and IL-13. In contrast, IL-21R-deficiency was associated with an increase in PBS- and allergen-driven IgE levels, while IgG1 and IgG2a levels were decreased. Moreover, our results suggest that IL-21 may contribute to AHR through its ability to both directly induce Th2 cell survival and to impair regulatory T-cell suppression of Th2 cytokine production. Importantly, we show that IL-21-positive cells are increased in the bronchial mucosa of asthmatics compared with non-asthmatics. These results suggest that IL-21 plays an important role in the allergic diathesis by enhancing Th2 cytokine production through multiple mechanisms including the suppression of Treg inhibitory effects on Th2 cell cytokine production.