Merck
  • Prognostic value of rubidium-82 positron emission tomography in patients after heart transplant.
Prognostic value of rubidium-82 positron emission tomography in patients after heart transplant.
Circulation. Cardiovascular imaging (2014-09-04)
Brian A Mc Ardle, Ross A Davies, Li Chen, Gary R Small, Terrence D Ruddy, Girish Dwivedi, Yeung Yam, Haissam Haddad, Lisa M Mielniczuk, Ellamae Stadnick, Renee Hessian, Ann Guo, Rob S Beanlands, Robert A deKemp, Benjamin J W Chow
ABSTRACT
Cardiac allograft vasculopathy is a key prognostic determinant after heart transplant. Detection and risk stratification of patients with cardiac allograft vasculopathy are problematic. Positron emission tomography using rubidium-82 allows quantification of absolute myocardial blood flow and may have utility for risk stratification in this population. Patients with a history of heart transplant undergoing dipyridamole rubidium-82 positron emission tomography were prospectively enrolled. Myocardial perfusion and left ventricular ejection fraction were recorded. Absolute flow quantification at rest and after dipyridamole stress as well as the ratio of mean global flow at stress and at rest, termed myocardial flow reserve, were calculated. Patients were followed for all-cause death, acute coronary syndrome, and heart failure hospitalization. A total of 140 patients (81% men; median age, 62 years; median follow-up, 18.2 months) were included. There were 14 events during follow-up (9 deaths, 1 acute coronary syndrome, and 4 heart failure admissions). In addition to baseline clinical variables (estimated glomerular filtration rate, previously documented cardiac allograft vasculopathy), relative perfusion defects, mean myocardial flow reserve, and mean stress myocardial blood flow were significant predictors of adverse outcome. Abnormalities on rubidium-82 positron emission tomography were predictors of adverse events in heart transplant patients. Larger prospective studies are required to confirm these findings.
Product Description
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Tantalum(V) ethoxide, packaged for use in deposition systems
Caffeine, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard