Merck
  • Chaotropic salts in liquid chromatographic method development for the determination of pramipexole and its impurities following quality-by-design principles.
Chaotropic salts in liquid chromatographic method development for the determination of pramipexole and its impurities following quality-by-design principles.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Ana Vemić, Tijana Rakić, Anđelija Malenović, Mirjana Medenica
ABSTRACT
The aim of this paper is to present a development of liquid chromatographic method when chaotropic salts are used as mobile phase additives following the QbD principles. The effect of critical process parameters (column chemistry, salt nature and concentration, acetonitrile content and column temperature) on the critical quality attributes (retention of the first and last eluting peak and separation of the critical peak pairs) was studied applying the design of experiments-design space methodology (DoE-DS). D-optimal design is chosen in order to simultaneously examine both categorical and numerical factors in minimal number of experiments. Two ways for the achievement of quality assurance were performed and compared. Namely, the uncertainty originating from the models was assessed by Monte Carlo simulations propagating the error equal to the variance of the model residuals and propagating the error originating from the model coefficients' calculation. The baseline separation of pramipexole and its five impurities is achieved fulfilling all the required criteria while the method validation proved its reliability.
Sigma-Aldrich
Benzothiazole, 96%
Sigma-Aldrich
Sodium perchlorate hydrate, 99.99% trace metals basis
Sigma-Aldrich
Benzothiazole, ≥96%, FG
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium trifluoroacetate, 98%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Sodium perchlorate monohydrate, ACS reagent
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Sodium perchlorate monohydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%