Merck
  • A dual-responsive fluorescence method for the detection of clenbuterol based on BSA-protected gold nanoclusters.
A dual-responsive fluorescence method for the detection of clenbuterol based on BSA-protected gold nanoclusters.
Analytica chimica acta (2015-04-08)
Xueling Cao, Hongwei Li, Lili Lian, Na Xu, Dawei Lou, Yuqing Wu
ABSTRACT
The illegal feeding of clenbuterol (CLB) to domestic animals and the potential harm of it to human health lead an urgent requirement for the efficient detection of CLB, especially in the edible meat. In this paper we reported a new fluorescence method for the detection of trace amount of CLB by using the BSA-protected gold nanoclusters (AuNCs@BSA). Under the excitation of either 280 or 500 nm the emission of AuNCs@BSA was quenched obviously by diazotized CLB, supplying a dual-responsive fluorescence method to detect CLB in aqueous solution. In addition, the linear response of the fluorescence intensity of AuNCs@BSA to diazotized CLB allowed the quantitative detection of CLB in a range of 4.0 nM-300 μM upon excitation at two wavelength, and the limit of detection for CLB was 3.0 nM upon 280 nm excitation and 1.6 nM upon 500 nm excitation, respectively. In addition, the dual-responsive mechanism of AuNCs@BSA to CLB was investigated in detail by using several CLB analogues and reference compounds. Particularly, the proposed method was successfully applied to detect CLB in pork mince and the results were validated well by HPLC, illustrating it could be used as a reliable, rapid, and cost-effective technique for the determination of CLB residues in real samples.
Product Description
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Supelco
Sodium nitrite, analytical standard