Merck
  • Efficacy of albendazole in combination with thymol against Echinococcus multilocularis protoscoleces and metacestodes.
Efficacy of albendazole in combination with thymol against Echinococcus multilocularis protoscoleces and metacestodes.
Acta tropica (2014-08-26)
Albani Clara María, Elissondo María Celina
ABSTRACT
The larval (metacestode) stage of the tapeworm Echinococcus multilocularis causes alveolar echinococcosis (AE), a mainly hepatic disease characterized by continuous asexual proliferation of metacestodes by exogenous budding, resulting in the tumor-like, infiltrative growth of the parasite lesion. Current chemotherapeutical treatment of AE relies on the use of benzimidazoles, albendazole (ABZ) and mebendazole, but these drugs act parasitostatic rather than parasitocidal, and due to their low success rate they imply a lifelong application causing severe side effects. Thymol is one of the major components of the essential oils of Thymus and is a widely known anti-microbial agent. The aim of the present work was to compare the efficacy of albendazole (ABZ) and thymol separately or combined on E. multilocularis protoscoleces and metacestodes. For this purpose, microscopical examinations at different time points were carried out. Moreover the tegumentary enzyme gamma glutamyl transferase (GGT) was measured to quantify the damage in metacestodes. Even though treatments of in vitro cultured E. multilocularis protoscoleces or metacestodes with ABZ or/and thymol showed that the drugs have an adverse effect on parasite viability, the combination of the two compounds at the concentration of 10μg/ml showed the maximum anti-parasitic effect. Three days postincubation the first effects of the treatment were detected on protoscoleces and a marked reduction in viability (33%) was registered at day 18. Incubation of E. multilocularis metacestodes in the presence of ABZ 10μg/ml+thymol 10μg/ml during 10 days resulted in dramatic alterations such as strongly irregular and fissured surface and markedly disrupted vesicles. Scanning electron microscopy showed that protoscoleces as well as the germinal layer of E. multilocularis metacestodes were dramatically damaged following ABZ or/and thymol treatment. Also an important increase of tegumentary enzyme GGT was registered after 72h postincubation with both drugs. The data reported in this article demonstrate a clear in vitro effect of ABZ+thymol against E. multilocularis protoscoleces and metacestodes.
Product Description
Supelco
Albendazole, analytical standard, ≥98%
USP
Albendazole, United States Pharmacopeia (USP) Reference Standard
Albendazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thymol, FCC, FG
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Thymol, analytical standard
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Thymol, ≥98.5%
Supelco
Thymol, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Thymol, European Pharmacopoeia (EP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard