Merck
  • A novel dereplication strategy for the identification of two new trace compounds in the extract of Gastrodia elata using UHPLC/Q-TOF-MS/MS.
A novel dereplication strategy for the identification of two new trace compounds in the extract of Gastrodia elata using UHPLC/Q-TOF-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-10)
Zhifeng Li, Yawei Wang, Hui Ouyang, Yu Lu, Yan Qiu, Yulin Feng, Hongliang Jiang, Xin Zhou, Shilin Yang
ABSTRACT
An ultra performance liquid chromatography (UHPLC) coupled with electrospray ionization quadrupole time-of-flight mass spectrometry (ESI-Q-TOF-MS/MS) was used in the structural determination of natural compounds in Gastrodia elata. A total of 64 compounds were identified or tentatively characterized. The strategy used for characterization was comparing their retention time and fragmentation behaviors with those of the reference standards, or investigating their accurate mass measurements and characteristic fragmentation patterns followed by low-energy collision dissociation tandem mass spectrometry (CID-MS/MS). Phenolic conjugates mainly underwent consecutive losses of gastrodin residues and combined losses of H2O and CO2 from their citric acid units under negative MS/MS conditions. According to these rules, we have successfully characterized fifteen potential novel compounds. To confirm the reliability of this strategy, two targeted unknown trace parishins were obtained from G. elata by LC/MS-guided isolation. Based on the analysis of data from NMR spectroscopy and other techniques, the two unknown parishins were identified as 2-[4-O-(β-d-glucopyranosyl)benzyl]-3-methyl-citrate (parishin J) and 1,2-di-[4-O-(β-d-glucopyranosyl)benzyl]-3-methyl-citrate (parishin K), respectively. The fully established structures were consistent with the MS-oriented structural elucidation. This study expanded our knowledge on parishins in Gastrodia species, and the proposed strategy was proven efficient and reliable in the discovery of new minor compounds from herbal extracts.
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard