Merck
  • PRL-3 mediates the protein maturation of ULBP2 by regulating the tyrosine phosphorylation of HSP60.
PRL-3 mediates the protein maturation of ULBP2 by regulating the tyrosine phosphorylation of HSP60.
Journal of immunology (Baltimore, Md. : 1950) (2015-02-18)
Wai-Hang Leung, Queenie P Vong, Wenwei Lin, David Bouck, Susanne Wendt, Erin Sullivan, Ying Li, Rafijul Bari, Taosheng Chen, Wing Leung
ABSTRACT
Many malignant cells release the NKG2D ligand ULBP2 from their cell surface to evade immunosurveillance by NK cells and CD8 T cells. Although the shedding mechanism remains unclear, various inhibitors of matrix metalloproteinases have been shown to efficiently block the release of soluble ULBP2. The clinical use of these inhibitors, however, is limited because of adverse side effects. Using high-throughput screening technique, we identified a specific inhibitor of phosphatase of regenerating liver 3 (PRL-3) that could reduce the level of soluble ULBP2 in the culture supernatant of various cancer cell lines. Inhibition or gene knockdown of PRL-3 did not reduce ULBP2 shedding, but rather suppressed posttranslational maturation of ULBP2, resulting in intracellular retention of immature ULBP2. We then found that ULBP2 was constitutively associated with heat shock protein HSP60. Complete maturation of ULBP2 required tyrosine phosphorylation of HSP60 which was mediated by PRL-3.
Product Description
SAFC
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
SAFC
BIS-TRIS
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase substrate, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
SKU
Pack Size
Availability
Price
Quantity
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Staurosporine, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)