Merck
  • Evaluation of serum phosphopeptides as potential cancer biomarkers by mass spectrometric absolute quantification.
Evaluation of serum phosphopeptides as potential cancer biomarkers by mass spectrometric absolute quantification.
Talanta (2014-05-21)
Guijin Zhai, Xiaoyan Wu, Qun Luo, Kui Wu, Yao Zhao, Jianan Liu, Shaoxiang Xiong, Yu-Qi Feng, Liping Yang, Fuyi Wang
ABSTRACT
Mass spectrometric quantification of phosphopeptides is a challenge due to the ion suppression effect of highly abundant non-phosphorylated peptides in complex samples such as serum. Several strategies for relative quantification of serum phosphopeptides based on MS have been developed, but the power of relative quantities was limited when making direct comparisons between two groups of samples or acting as a clinical examination index. Herein, we describe an MS absolute quantification strategy combined with Titania Coated Magnetic Hollow Mesoporous Silica Microspheres (TiO2/MHMSM) enrichment and stable isotopic acetyl labeling for phosphopeptides in human serum. Four endogenous serum phosphopeptides generated by degradation of fibrinogen were identified by LC-ESI-MS/MS following TiO2/MHMSM enrichment. The ESI-MS signal intensity ratios of the four phosphopeptide standards labeled with N-acetoxy-H3-succinimide (H3-NAS) and N-acetoxy-D3-succinimide (D3-NAS), following TiO2/MHMSM capture are linearly correlated with the molar ratios of the "light" to "heavy" phosphopeptides over the range of 0.1-4 with an r(2) of up to 0.998 and a slope of close to 1. The recovery of the four phosphopeptides spiked at low, medium and high levels in human sera were 98.4-111.9% with RSDs ranging 2.0-10.1%. The absolute quantification of the phosphopeptides in serum samples of 20 healthy persons and 20 gastric cancer patients by the developed method demonstrated that 3 out of the 4 phosphopeptides showed remarkable variation in serum level between healthy and cancer groups, and the phosphopeptide DpSGEGDFLAEGGGVR is significantly down-regulated in the serum of patients, being a potential biomarker for gastric cancer diagnosis.
Sigma-Aldrich
N-Acetoxy-d3-succinimide, 98 atom % D
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%