Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Pricing and availability is not currently available.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Pricing and availability is not currently available.
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Pricing and availability is not currently available.
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Fast Blue B Salt, Dye content ~95 %
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediamine, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
5(6)-Carboxyfluorescein, Dye content 90 %
Pricing and availability is not currently available.
Sigma-Aldrich
5(6)-Carboxyfluorescein, suitable for fluorescence, BioReagent, ≥95% (HPLC)
Pricing and availability is not currently available.
Supelco
Ethylenediamine, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Pricing and availability is not currently available.