Merck
  • Empowering a transition-metal-free coupling between alkyne and alkyl iodide with light in water.
Empowering a transition-metal-free coupling between alkyne and alkyl iodide with light in water.
Nature communications (2015-03-11)
Wenbo Liu, Lu Li, Chao-Jun Li
ABSTRACT
Methods to assemble alkynes are essential for synthesizing fine chemicals, pharmaceuticals and polymeric photo-/electronic materials. Using light as a clean energy form and water as a green solvent has the potential to make synthetic chemistry more environmentally friendly. Here we present a transition-metal-free coupling protocol between aryl alkyne and alkyl iodide enabled by photoenergy in water. Under ultraviolet irradiation and in basic aqueous media, aryl alkynes efficiently couple with a wide range of alkyl iodides including primary, secondary and tertiary ones under mild conditions. A tentative mechanism for the coupling is also proposed.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%