Merck
  • Direct detection of Mycobacterium tuberculosis in sputum using combined solid phase extraction-gas chromatography-mass spectrometry.
Direct detection of Mycobacterium tuberculosis in sputum using combined solid phase extraction-gas chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-03)
Ngoc A Dang, Marta Mourão, Sjoukje Kuijper, Elisabetta Walters, Hans-Gerd Janssen, Arend H J Kolk
ABSTRACT
Recently, thermally-assisted hydrolysis and methylation followed by gas chromatography-mass spectrometry (THM-GC-MS) in combination with chemometrics has been used to develop a 20-compound model for fast differentiation of Mycobacterium tuberculosis (MTB) from Non-tuberculous mycobacteria (NTM) in bacterial cultures. This model provided better than 95% accuracy. In our current work a hexane/methanol/water extraction followed by a solid phase extraction (SPE) clean-up procedure was developed for use before THM-GC-MS, to make the test suitable for the identification of mycobacteria in sputum. The 20 biomarker model had to be adapted since many compounds were also found in the sputum of non-tuberculosis patients. An algorithm was established based on tuberculostearic acid, hexacosanoic acid and mycoserosates. The detection limit of the method was approximately 1×10(4) bacteria/mL sputum. Sputum specimens from 32 patients from South Africa who were suspected of having tuberculosis were blindly tested using the new method. Eight of the nine culture-positive sputum specimens were detected by the new SPE-THM-GC-MS method, resulting in a sensitivity of 89%. The specimen that was missed by the new method was also microscopy negative. The specificity of the test was 100%; all 23 microscopy- and culture-negative specimens were correctly identified as negative by SPE-THM-GC-MS.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%