Merck
  • Butachlor causes disruption of HPG and HPT axes in adult female rare minnow (Gobiocypris rarus).
Butachlor causes disruption of HPG and HPT axes in adult female rare minnow (Gobiocypris rarus).
Chemico-biological interactions (2014-08-12)
Lifei Zhu, Wei Li, Jinmiao Zha, Miao Wang, Lilai Yuan, Zijian Wang
ABSTRACT
Butachlor is a chloroacetamide herbicide widely used in Asia, and may enter the aquatic environment through agricultural application. In this study, plasma VTG and hormone levels (E2, 11-KT, T3 and T4) were determined after the female rare minnow (Gobiocypris rarus) was exposed to butachlor at environmental relevant concentrations (0, 0.1, 1, and 10μg/L) for 40days. The mRNA levels of the HPG axis-related genes (gnrh, erα, vtg, star, lhr, 3β-hsd, cyp11a, cyp17, cyp19a and cyp19b), and the HPT axis-related genes (trα, dio1, dio2, and dio3) were quantified after 20 and 40days exposure to butachlor. For the HPG axis, the plasma 11-KT was increased at exposure concentration of 10μg/L, and VTG was significantly decreased at 1μg/L. Functional genes like gnrh and cyp19b in the brains, star, lhr, cyp11a, 3β-hsd, and cyp19a in the ovaries, and erα and vtg in livers were up-regulated. For the HPT axis, the results showed that plasma T4 levels were significantly increased, the gene expression of dio1 was up-regulated, dio2 showed no significant variation, and dio3 was down-regulated in the livers. These results indicated that butachlor may promote the accumulation of T4 in fish through inactive deiodinase type 3. The transcription of HPG axis-related genes could serve as an auto-regulation of hormone levels after exposure to butachlor. Furthermore, the activation of gnrh may play an important role as a feed-back mechanism in the regulation of hormone levels and crosstalk of endocrine axes.
Product Description
Supelco
Butachlor, PESTANAL®, analytical standard
Pricing and availability is not currently available.