Merck
  • MicroRNA-1 functions as a potential tumor suppressor in osteosarcoma by targeting Med1 and Med31.
MicroRNA-1 functions as a potential tumor suppressor in osteosarcoma by targeting Med1 and Med31.
Oncology reports (2014-06-28)
Chaoyin Jiang, Hua Chen, Lei Shao, Qiaojie Wang
ABSTRACT
MicroRNA-1 (miR-1) has been shown to function as a critical gene regulator in multiple types of cancers. However, the role of miR-1 in osteosarcoma has not been totally clarified. In the present study, we investigated the effects of miR-1 on osteosarcoma and the underlying mechanism. We found that miR-1 was downregulated in osteosarcoma tissues and osteosarcoma cell lines. Restoration of miR-1 significantly suppressed osteosarcoma cell proliferation by inhibiting cell cycle progression. Mediator complex subunit 1 (Med1) and 31 (Med31) were validated as targets of miR-1 in osteosarcoma by luciferase reporter assay. Downregulation of Med1 and Med31 suppressed the proliferation of osteosarcoma cells, and overexpression of Med1 and Med31 abrogated the effects of miR-1 on cell proliferation. Furthermore, both miR-1 and knockdown of Med1 or Med31 reduced the expression of met proto-oncogene (MET) and blocked the downstream signaling of MET responding to hepatocyte growth factor (HGF). Taken together, the findings of this study suggest that Med1 and Med31 serve as potential gene therapeutic targets in osteosarcoma and miR-1 may prove to be a promising agent.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Med31
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
MISSION® esiRNA, targeting human MED31