Merck
  • Seasonal phytochemical study and antimicrobial potential of Vetiveria zizanioides roots.
Seasonal phytochemical study and antimicrobial potential of Vetiveria zizanioides roots.
Acta pharmaceutica (Zagreb, Croatia) (2014-12-23)
Delair Silveira Dos Santos, Júlia Vieira Oberger, Rivaldo Niero, Theodoro Wagner, Franco Delle Monache, Alexandre Bella Cruz, Zhelmy Martin-Quintal, Valdir Cechinel Filho
ABSTRACT
This paper describes the seasonal phytochemical variation and the antimicrobial potential of V. zizanioides roots collected in Brazil. Considering the high levels of chemical constituents and their biological activity in dichloromethane fraction, the plants were grown in different seasons and the respective dichloromethane fractions were analyzed by gas chromatography-mass spectrometry. The antimicrobial activity was evaluated against several pathogenic microorganisms by determining the minimum inhibitory concentration (MIC) using the agar dilution method. Yields of dichloromethane fractions from plants collected in the autumn and spring occurred in a higher proportion than in other seasons. Khusimol (2) was isolated by column chromatography and identified by NMR and CG-MS, along with other sesquiterpenes, including β-vetivenene (1), vetiselinenol (3), isovalencenol (4), vetivenic acid (5), α-vetivone (6) and β-vetivone (7). Some extracts showed promising antimicrobial effects, with MICs ranging from 31.25 to 500 μg mL-1. Kushimol was slightly active against the tested microorganisms.
Product Description
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material