Merck
  • Ultra-performance liquid chromatographic determination of tocopherols and retinol in human plasma.
Ultra-performance liquid chromatographic determination of tocopherols and retinol in human plasma.
Journal of chromatographic science (2013-10-31)
Edward C Bell, Mathew John, Rodney J Hughes, Thu Pham
ABSTRACT
A rapid, selective and sensitive ultra-performance liquid chromatography method has been developed for the detection and quantification of tocopherols and retinol in human plasma. Alpha-tocopherol, gamma-tocopherol and retinol are assayed using fluorescence detection. Excitation/emission wavelengths are 295/330 nm and 325/470 nm for the analysis of both tocopherols and retinol, respectively. Retinol acetate is employed as the internal standard. The reversed-phase method incorporates gradient elution with a mobile phase consisting of methanol and acetonitrile. Separation of vitamin compounds is achieved using a bridged ethyl hybrid C18 column. The retention times for retinol, retinol acetate, gamma-tocopherol and alpha-tocopherol are 1.6, 1.8, 3.9 and 4.3 min, respectively. The limits of quantification for retinol, gamma-tocopherol and alpha-tocopherol were 0.02, 0.02 and 0.1 µg/mL, respectively. The assay method is suitable for the analysis of tocopherols and retinol in human plasma. The method may be applied following the ingestion of foods fortified with these fat-soluble vitamins.
Product Description
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%