Merck
  • Synthesis of a new molecularly imprinted polymer for sorption of the silver ions from geological and antiseptic samples for determination by flame atomic absorption spectrometry.
Synthesis of a new molecularly imprinted polymer for sorption of the silver ions from geological and antiseptic samples for determination by flame atomic absorption spectrometry.
Journal of AOAC International (2015-04-24)
Hamid Hashemi-Moghaddam, Faegheh Yahyazadeh, Mohammad Taghi Vardini
ABSTRACT
A new molecularly imprinted polymer (MIP) was synthesized using methacrylic acid (functional monomer), ethylene glycol dimethacrylate (crosslinker), 2,2'-azobisisobutironitril (initiator), silver (Ag) dithizone complex (template), and chloroform (porogenic solvent). This process was a noncovalent, bulk, thermal radical-polymerization. To compare the performance of this polymer, control polymer (nonimprinted polymer) was prepared under well-defined conditions without the use of a template. Extraction experiments were performed on the MIP and a nonimprinted polymer. Then, various parameters were optimized, such as pH, time, concentration of sample, and type of eluent for elution of Ag from polymer. In addition, interfering effects were investigated on the absorption of Ag by the MIP. This polymer was used for the rapid extraction and preconcentration of Ag from an antiseptic and geological sample. Finally, the amount of Ag was measured by flame atomic absorption spectrometry after preconcentration by the synthesized MIP, and results were compared with a direct inductively coupled plasma method. The results showed high performance of this method in preconcentration of Ag.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material