Merck
  • Detoxification of biomass hydrolysates with nucleophilic amino acids enhances alcoholic fermentation.
Detoxification of biomass hydrolysates with nucleophilic amino acids enhances alcoholic fermentation.
Bioresource technology (2015-03-31)
Rui Xie, Maobing Tu, Jamarius Carvin, Yonnie Wu
ABSTRACT
Carbonyl compounds generated in biomass pretreatment hinder the biochemical conversion of biomass hydrolysates to biofuels. A novel approach of detoxifying hydrolysates with amino acids for ethanol production was developed. Among the 20 amino acids assessed for their detoxification efficiency and nucleophilicity, cysteine was the most effective one. It increased both ethanol productivity and final yield of biomass hydrolysates from 0.18 (untreated) to 1.77 g/L/h and from 0.02 to 0.42 g/g, respectively. Detoxification efficiency was followed by histidine and it increased the final yield to 0.42 g/g, then by lysine, tryptophan and asparagine. It was observed all five effective amino acids contained reactive side-chain functional groups, which played important roles in the amino acid detoxification reaction. The study further showed cysteine and glycine detoxifications were temperature and pH dependent. The mechanistic study using mass spectrometry revealed thiazolidine carboxylic acid, a Schiff base, was formed by condensation of aldehyde and cysteine.
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Levulinic acid, ≥97%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG
Supelco
Acetic acid, analytical standard
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
Levulinic acid, 98%
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material