Merck
  • HN1 negatively influences the β-catenin/E-cadherin interaction, and contributes to migration in prostate cells.
HN1 negatively influences the β-catenin/E-cadherin interaction, and contributes to migration in prostate cells.
Journal of cellular biochemistry (2014-08-30)
Lokman Varisli, Bilge E Ozturk, Gencer K Akyuz, Kemal S Korkmaz
ABSTRACT
Previously, it has been reported that HN1 is involved in cytoplasmic retention and degradation of androgen receptor in an AKT dependent manner. As HN1 is a hormone inducible gene, and has been shown that it is upregulated in various cancers, we studied the importance of HN1 function in β-catenin signaling in prostate cancer cell line, PC-3 and mammary cancer cell line MDA-MB231. Here, we demonstrated that HN1 physically associates with GSK3β/β-catenin destruction complex and abundantly localizes to cytoplasm, especially when the GSK3β is phosphorylated on S9 residue. Further, ectopic HN1 expression results an increase in the β-catenin degradation leading to loss of E-cadherin interaction, concurrently contributing to actin re-organization, colony formation and migration in cancer cell lines. Thus, we report that HN1 is an essential factor for β-catenin turnover and signaling, augments cell growth and migration in prostate cancer cells.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hn1
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human HN1