Merck
  • Caffeic acid, naringenin and quercetin enhance glucose-stimulated insulin secretion and glucose sensitivity in INS-1E cells.
Caffeic acid, naringenin and quercetin enhance glucose-stimulated insulin secretion and glucose sensitivity in INS-1E cells.
Diabetes, obesity & metabolism (2013-11-12)
S Bhattacharya, N Oksbjerg, J F Young, P B Jeppesen
ABSTRACT
Caffeic acid, naringenin and quercetin are naturally occurring phenolic compounds (PCs) present in many plants as secondary metabolites. The aim of this study was to investigate their effect on glucose-stimulated insulin secretion (GSIS) in INS-1E cells and to explore their effect on expression of genes involved in β-cell survival and function under normoglycaemic and glucotoxic conditions. For acute studies, INS-1E cells were grown in 11 mM glucose (72 h) and then incubated with the PCs (1 h) with 3.3/16.7 mM glucose; whereas, for chronic studies, the cells were grown in 11 mM glucose (72 h) with/without the PCs, and then incubated with 3.3/16.7 mM glucose (1 h); thereafter, GSIS was measured. For GSIS and gene expression studies (GES) under glucotoxic conditions, two sets of cells were grown in 11/25 mM glucose with/without the PCs (72 h): one was used for GES, using real time RT-PCR, and the other was exposed to 3.3/16.7 mM glucose, followed by measurement of GSIS. The study demonstrated that the PCs can enhance GSIS under hyperglycaemic and glucotoxic conditions in INS-1E cells. Moreover, these compounds can differentially, yet distinctly change the expression profile of genes [Glut2 (glucose transporter 2), Gck (glucokinase), Ins1 (insulin 1), Ins2, Beta2 (neurogenic differentiation protein 1), Pdx1 (pancreatic and duodenal homeobox protein 1), Akt1 (RAC-α serine/threonine-protein kinase encoding gene), Akt2 (RAC-β serine/threonine-protein kinase encoding gene), Irs1 (insulin receptor substrate 1), Acc1 (acetyl CoA carboxylase 1), Bcl2 (β-cell lymphoma 2 protein), Bax (Bcl-2 associated X protein), Casp3 (Caspase 3), Hsp70 (heat shock protein 70), and Hsp90] involved in β-cell stress, survival and function. The results indicate that the PCs tested enhance GSIS and glucose sensitivity in INS-1E cells. They also modulate gene expression profiles to improve β-cell survival and function during glucotoxicity.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
(±)-Naringenin, analytical standard
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)