Merck
  • Tmem27 is upregulated by vitamin D in INS-1 cells and its serum concentrations are low in patients with autoimmune diabetes.
Tmem27 is upregulated by vitamin D in INS-1 cells and its serum concentrations are low in patients with autoimmune diabetes.
Scandinavian journal of clinical and laboratory investigation (2014-04-04)
Milaim Pepaj, Nina Gjerlaugsen, Kari Julien, Per M Thorsby
ABSTRACT
Transmembrane protein 27 (Tmem27), which is expressed in pancreatic β-cells, plays an important role in insulin secretion and pancreatic β-cell proliferation. Analysis of the INS-1 cell proteome using stable isotope labeling by amino acids in cell culture (SILAC) in combination with LC-MS identified Tmem27 as the one of most robustly (up to seven-fold) upregulated proteins after treatment with the active metabolite of vitamin D, 1,25-(OH)2D3. Furthermore, we report that Tmem27 which is cleaved and released from, i.e. pancreatic β-cells, is present in human serum and its levels are significantly lower in subjects with autoimmune diabetes as compared to healthy individuals (13% of the levels). Additionally, Tmem27 correlated positively (0.70) with C-peptide serum levels in healthy subjects. Our data indicate that Tmem27 could be of potential value as a serum marker for the pathogenesis of diabetes and as such may warrant the development of measurement methods with lower limit of detection for its further validation.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard