Merck
  • Risk assessment of pesticides detected in surface water of the Alqueva reservoir (Guadiana basin, southern of Portugal).
Risk assessment of pesticides detected in surface water of the Alqueva reservoir (Guadiana basin, southern of Portugal).
The Science of the total environment (2014-05-20)
P Palma, M Köck-Schulmeyer, P Alvarenga, L Ledo, I R Barbosa, M López de Alda, D Barceló
ABSTRACT
The purpose of this study was to evaluate the potential impact of the pesticides detected in the Alqueva reservoir (Guadiana Basin, South Iberian Peninsula) on the aquatic organisms belonging to this ecosystem. For this purpose, the occurrence and risk assessment of 25 pesticides and of a number of their degradation products were determined in the Alqueva surface waters. The areas (sampling stations) most polluted by pesticides were Sra. Ajuda, Lucefecit and Álamos in the northern and in the middle portions of the reservoir, respectively. The aquatic risk assessment revealed that from the various compounds analysed terbuthylazine, chlorfenvinphos and diazinon presented non-acceptable risk when maximum concentrations were used as the measured environmental concentrations (MEC). The locations that had more samples with risk quotients higher than 1 (high risk) were Sra. Ajuda followed by Lucefécit. The use of risk assessment allowed us to conclude that, despite that the pesticides' concentrations in the water column fulfil the European environmental quality standards, a number of the compounds show a high ecotoxicological risk for the aquatic organisms in the Alqueva ecosystem. The results thereby demonstrate that to have an efficient risk management process, the regulatory authorities of each country must consider an integrative chemical and ecotoxicological approach.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
Linuron, PESTANAL®, analytical standard
Supelco
Malathion, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
Supelco
Atrazine-desisopropyl, PESTANAL®, analytical standard
Supelco
Isoproturon, PESTANAL®, analytical standard
Supelco
Diuron, PESTANAL®, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Dimethoate, PESTANAL®, analytical standard
Supelco
Malaoxon, PESTANAL®, analytical standard
Supelco
Propanil, PESTANAL®, analytical standard
Supelco
Chlorfenvinphos, PESTANAL®, analytical standard
Supelco
Fenitrothion, PESTANAL®, analytical standard
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Malathion, certified reference material, TraceCERT®
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Malathion, European Pharmacopoeia (EP) Reference Standard
USP
Malathion, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG