Merck
  • Efficient HPLC method development using structure-based database search, physico-chemical prediction and chromatographic simulation.
Efficient HPLC method development using structure-based database search, physico-chemical prediction and chromatographic simulation.
Journal of pharmaceutical and biomedical analysis (2014-12-08)
Lin Wang, Jinjian Zheng, Xiaoyi Gong, Robert Hartman, Vincent Antonucci
ABSTRACT
Development of a robust HPLC method for pharmaceutical analysis can be very challenging and time-consuming. In our laboratory, we have developed a new workflow leveraging ACD/Labs software tools to improve the performance of HPLC method development. First, we established ACD-based analytical method databases that can be searched by chemical structure similarity. By taking advantage of the existing knowledge of HPLC methods archived in the databases, one can find a good starting point for HPLC method development, or even reuse an existing method as is for a new project. Second, we used the software to predict compound physicochemical properties before running actual experiments to help select appropriate method conditions for targeted screening experiments. Finally, after selecting stationary and mobile phases, we used modeling software to simulate chromatographic separations for optimized temperature and gradient program. The optimized new method was then uploaded to internal databases as knowledge available to assist future method development efforts. Routine implementation of such standardized workflows has the potential to reduce the number of experiments required for method development and facilitate systematic and efficient development of faster, greener and more robust methods leading to greater productivity. In this article, we used Loratadine method development as an example to demonstrate efficient method development using this new workflow.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Loratadine, ≥98% (HPLC), powder
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Supelco
Loratadine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M