Merck
  • A low-frequency variant in MAPK14 provides mechanistic evidence of a link with myeloperoxidase: a prognostic cardiovascular risk marker.
A low-frequency variant in MAPK14 provides mechanistic evidence of a link with myeloperoxidase: a prognostic cardiovascular risk marker.
Journal of the American Heart Association (2014-08-29)
Dawn M Waterworth, Li Li, Robert Scott, Liling Warren, Christopher Gillson, Jennifer Aponte, Lea Sarov-Blat, Dennis Sprecher, Josée Dupuis, Alex Reiner, Bruce M Psaty, Russell P Tracy, Honghuang Lin, Ruth McPherson, Stephanie Chissoe, Nick Wareham, Margaret G Ehm
ABSTRACT
Genetics can be used to predict drug effects and generate hypotheses around alternative indications. To support Losmapimod, a p38 mitogen-activated protein kinase inhibitor in development for acute coronary syndrome, we characterized gene variation in MAPK11/14 genes by exome sequencing and follow-up genotyping or imputation in participants well-phenotyped for cardiovascular and metabolic traits. Investigation of genetic variation in MAPK11 and MAPK14 genes using additive genetic models in linear or logistic regression with cardiovascular, metabolic, and biomarker phenotypes highlighted an association of RS2859144 in MAPK14 with myeloperoxidase in a dyslipidemic population (Genetic Epidemiology of Metabolic Syndrome Study), P=2.3×10(-6)). This variant (or proxy) was consistently associated with myeloperoxidase in the Framingham Heart Study and Cardiovascular Health Study studies (replication meta-P=0.003), leading to a meta-P value of 9.96×10(-7) in the 3 dyslipidemic groups. The variant or its proxy was then profiled in additional population-based cohorts (up to a total of 58 930 subjects) including Cohorte Lausannoise, Ely, Fenland, European Prospective Investigation of Cancer, London Life Sciences Prospective Population Study, and the Genetics of Obesity Associations study obesity case-control for up to 40 cardiovascular and metabolic traits. Overall analysis identified the same single nucleotide polymorphisms to be nominally associated consistently with glomerular filtration rate (P=0.002) and risk of obesity (body mass index ≥30 kg/m(2), P=0.004). As myeloperoxidase is a prognostic marker of coronary events, the MAPK14 variant may provide a mechanistic link between p38 map kinase and these events, providing information consistent with current indication of Losmapimod for acute coronary syndrome. If replicated, the association with glomerular filtration rate, along with previous biological findings, also provides support for kidney diseases as alternative indications.