Merck
  • Study of chemical composition and volatile compounds along with in-vitro assay of antioxidant activity of two medicinal rice varieties: Karungkuravai and Mappilai samba.
Study of chemical composition and volatile compounds along with in-vitro assay of antioxidant activity of two medicinal rice varieties: Karungkuravai and Mappilai samba.
Journal of food science and technology (2015-04-22)
K Krishnanunni, P Senthilvel, Sudha Ramaiah, Anand Anbarasu
ABSTRACT
The objective of this study is to analyze the chemical composition and volatiles present in two medicinal rice varieties of "karungkuravai" and "mappilai samba" and to investigate their total phenolic content and bio activity. Chemical composition of the rice varieties was analyzed using gas chromatography/mass spectroscopy (GC-MS), the volatile compounds are identified using static head space analysis (SHS) followed by GC-MS. Total phenolic content (TPC) is estimated using Folin Ciocalteu colorimetric method, antioxidant assay is done using 1, 1-diphenyl-2-picrylhydrazyl (DPPH) assay and ferric reducing antioxidant power (FRAP) assay. GC-MS and SHS analysis identified pharmaceutically important phytochemicals present in the rice variety. The compounds like "curlone" and "7-chloro-1,3-dihydro-5-phenyl-1-(trimethylsilyl)- 2H-1-4-benzodiazapene-2-one" are identified for the first time in any rice variety. Phenols are found to be present in both rice varieties. Both rice varieties also showed antioxidant activity in both DPPH and FRAP assays and the IC50 values were 91.08 μg/ml and 359.93 μg/ml for "karungkuravai" and "mappilai samba", respectively. . The correlation coefficient and regression analysis of total phenolic content with DPPH assay and FRAP assay show significant positive correlation coefficient values and coefficient of regression values.
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%