Merck
  • Scalable high-fidelity growth of semiconductor nanorod arrays with controlled geometry for photovoltaic devices using block copolymers.
Scalable high-fidelity growth of semiconductor nanorod arrays with controlled geometry for photovoltaic devices using block copolymers.
Small (Weinheim an der Bergstrasse, Germany) (2014-07-26)
Candice I Pelligra, Su Huang, Jonathan P Singer, Anthony T Mayo, Richard R Mu, Chinedum O Osuji
ABSTRACT
Controlled density semiconducting oxide arrays are highly desirable for matching nanometer length scales specific to emerging applications. This work demonstrates a facile one-step method for templating hydrothermal growth which provides arrays with high-fidelity tuning of nanorod spacing and diameter. This solution-based method leverages the selective swelling of block copolymer micelle templates, which can be rationally designed by tuning molecular weight and volume fraction.
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Supelco
2-Methoxyethanol, analytical standard
Sigma-Aldrich
Ethanolamine, ≥99%
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Aucubin, analytical standard
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥98%
Aucubin, primary reference standard
Sigma-Aldrich
2-Methoxyethanol, suitable for HPLC, ≥99.9%
Supelco
2-Methoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Methoxyethanol, contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%
Sigma-Aldrich
2-Methoxyethanol, ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard