Merck
  • Analysis of trace organic pollutants in wastewater to assess biodegradation using wrong-way-round ionization in liquid chromatography/tandem mass spectrometry.
Analysis of trace organic pollutants in wastewater to assess biodegradation using wrong-way-round ionization in liquid chromatography/tandem mass spectrometry.
Rapid communications in mass spectrometry : RCM (2014-04-25)
Lijuan Su, Wendell O Khunjar, Diana S Aga
ABSTRACT
Monitoring the concentrations of pharmaceuticals and personal care products (PPCPs) in wastewater is an integral step toward understanding the fate of these contaminants in wastewater treatment plants (WWTPs). This paper aims to develop a method that allows for the simultaneous analysis of multiple classes of PPCPs that can be used as tracers to assess the performance of WWTPs. Five PPCP tracers - carbamazepine (CBZ), 17α-ethinylestradiol (EE2), nonylphenol (NP), salicylic acid (SA), and trimethoprim (TMP) - were analyzed by liquid chromatography/triple quadrupole mass spectrometry (LC/MS/MS) using a highly basic mobile phase (pH 10.3). Conventionally, TMP (pKa 7.12) and CBZ (pKa 13.94) are analyzed in positive ion mode using an acidic mobile phase. However, the high pH mobile phase allowed the quantification of all the tracers by polarity switching, with TMP undergoing wrong-way-round (WWR) ionization. The instrument limits of detection for the five tracers, without solid-phase extraction, were in the range of 1.3 to 5.9 ng/mL, except for NP, which was 238 ng/mL. The signal-to-noise (S/N) ratios for TMP and CBZ with the mobile phase at pH 10.3 were higher than the S/N ratios observed at pH 2.7 under positive electrospray ionization. The mechanism of WWR ionization for TMP was investigated, and we propose that a charge transfer from solvent clusters to TMP molecules due to electrolytic reactions at the surface of the droplet leads to WWR ionization in electrospray. A method to simultaneously analyze five representative PPCP tracers with a wide range of pKa values using WWR ionization in LC/MS/MS with polarity switching was developed. The method was successfully used to monitor the selected PPCPs in samples from full-scale WWTPs to assess their biodegradation under various treatment conditions.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free