Merck
  • Optimization of combinational intranasal drug delivery system for the management of migraine by using statistical design.
Optimization of combinational intranasal drug delivery system for the management of migraine by using statistical design.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-02-14)
Animesh Kumar, Tarun Garg, Ganti S Sarma, Goutam Rath, Amit Kumar Goyal
ABSTRACT
Migraine is a chronic disorder characterized by significant headache and various associated symptoms which worsen with exertion. Zolmitriptan approved for use in the acute treatment of migraine and related vascular headaches but are limited by high pain recurrence due to rapid drug elimination. Combinationalformulationof triptans and a nonsteroidal anti-inflammatory drug may provide a quicker and longer duration of relief from the subsequent pain during the attack. In this study, we formulate a Zolmitriptan (ZT) & ketorolac tromethamine (KT) loaded thermo reversible in-situ mucoadhesive intranasal gel (TMISG) formulation which gels at the nasal mucosal temperature and contains a bioadhesive polymer (Xyloglucan) that lengthens the residence time will enhance the bioavailability of the combinational drugs. This study uses Box-Behnken design for the first time to develop, optimize the TMISG and assess factors affecting the critical quality attributes. Histopathological study of the nasal mucosa suggested that the formulation was safe for nasal administration. The statistical difference in absolute bioavailability between oral and intranasal route suggested that intranasal route had almost 21% increases in bioavailability for ZT and for KT there was 16% increase over oral formulations. Optimized formulation would help mitigate migraine associated symptoms much better over the currently available formulations.
Sigma-Aldrich
Zolmitriptan, ≥98% (HPLC)
Ketorolac trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Supelco
Ketorolac Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketorolac Tromethamine, United States Pharmacopeia (USP) Reference Standard
Ketorolac trometamol for peak identification, European Pharmacopoeia (EP) Reference Standard