Merck
  • Impairment of high-density lipoprotein resistance to lipid peroxidation and adipose tissue inflammation in obesity complicated by obstructive sleep apnea.
Impairment of high-density lipoprotein resistance to lipid peroxidation and adipose tissue inflammation in obesity complicated by obstructive sleep apnea.
The Journal of clinical endocrinology and metabolism (2014-05-16)
Rahul Yadav, Michael France, Reza Aghamohammadzadeh, Yifen Liu, Salam Hama, See Kwok, Jonathan Schofield, Peter Turkington, Akheel A Syed, Rayaz Malik, Philip Pemberton, Adam Greenstein, Paul Durrington, Basil Ammori, Martin Gibson, Maria Jeziorska, Handrean Soran
ABSTRACT
Obstructive sleep apnea (OSA) complicates morbid obesity and is associated with increased cardiovascular disease incidence. An increase in the circulating markers of chronic inflammation and dysfunctional high-density lipoprotein (HDL) occur in severe obesity. The objective of the study was to establish whether the effects of obesity on inflammation and HDL dysfunction are more marked when complicated by OSA. Morbidly obese patients (n = 41) were divided into those whose apnea-hypoapnea index (AHI) was more or less than the median value and on the presence of OSA [OSA and no OSA (nOSA) groups]. We studied the antioxidant function of HDL and measured serum paraoxonase 1 (PON1) activity, TNFα, and intercellular adhesion molecule 1 (ICAM-1) levels in these patients. In a subset of 19 patients, we immunostained gluteal sc adipose tissue (SAT) for TNFα, macrophages, and measured adipocyte size. HDL lipid peroxide levels were higher and serum PON1 activity was lower in the high AHI group vs the low AHI group (P < .05 and P < .0001, respectively) and in the OSA group vs the nOSA group (P = .005 and P < .05, respectively). Serum TNFα and ICAM-1 levels and TNFα immunostaining in SAT increased with the severity of OSA. Serum PON1 activity was inversely correlated with AHI (r = -0.41, P < .03) in the OSA group. TNFα expression in SAT directly correlated with AHI (r = 0.53, P < .03) in the subset of 19 patients from whom a biopsy was obtained. Increased serum TNFα, ICAM-1, and TNFα expression in SAT provide a mechanistic basis for enhanced inflammation in patients with OSA. Decreased serum PON1 activity, impaired HDL antioxidant function, and increased adipose tissue inflammation in these patients could be a mechanism for HDL and endothelial dysfunction.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)