Merck
  • MiR-223 promotes the cisplatin resistance of human gastric cancer cells via regulating cell cycle by targeting FBXW7.
MiR-223 promotes the cisplatin resistance of human gastric cancer cells via regulating cell cycle by targeting FBXW7.
Journal of experimental & clinical cancer research : CR (2015-04-19)
Xiaoying Zhou, Wujuan Jin, Hongyan Jia, Jin Yan, Guoxin Zhang
ABSTRACT
Increasing evidence showed that miRNAs serve as modulators of human cancer, either as oncogene or tumor suppressors. Cisplatin resistance is the most common cause of chemotherapy failure in gastric cancer (GC). However, the roles of miRNAs in cisplatin resistance of GC remain largely unknown. The aim of the study was to identify a novel miRNA/gene pathway that regulates the sensitivity of GC cells to cisplatin. In this study, we chose miR-223 by qRT-PCR analysis, the most significantly up-regulated miRNA in GC, to investigate its formation of DDP-resistant phenotype of GC cells and possible molecular mechanisms. We found that miR-223 was most significantly up-regulated miRNA in DDP-resistant GC cells compared with parental GC cells. Besides, its expression was also significantly up-regulated in GC tissues. FBXW7 was identified as the direct and functional target gene of miR-223. Overexpression of FBXW7 could mimic the effect of miR-223 down-regulation and silencing of FBXW7 could partially reverse the effect of miR-223 down-regulation on DDP resistance of DDP-resistant GC cells. Besides, miR-223 and FBXW7 could affect the G1/S transition of cell cycle by altering some certain cell cycle regulators. Furthermore, miR-223 was found to be significantly up-regulated in H. pylori infected tissues and cells, suggesting that H. pylori infection may lead to GC development and DDP resistance. Our findings revealed the roles of miR-223/FBXW7 signaling in the DDP resistance of GC cells and targeting it will be a potential strategic approach for reversing the DDP resistance in human GC.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human FBXW7
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fbxw7
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%