Merck
  • Carbon-14 radiolabeling and in vivo biodistribution of a potential anti-TB compound.
Carbon-14 radiolabeling and in vivo biodistribution of a potential anti-TB compound.
Journal of labelled compounds & radiopharmaceuticals (2015-02-19)
Molahlehi S Sonopo, Kobus Venter, Grant Boyle, Susan Winks, Biljana Marjanovic-Painter, Jan R Zeevaart
ABSTRACT
A potential anti-TB compound bearing a nitroimidazole moiety from iThemba Pharmaceuticals TB chemical library exhibits promising in vitro activity in the microplate almar blue assay (MABA) with a minimum inhibitory concentration (MIC) value of 3 µg/mL. It is equipotent to the front-line drug Isoniazid, but the compound is less toxic with an IC50 of >100 µg/mL. Therefore, this potential iThemba nitroimidazole, 4-([1,1'-[(14)C6]biphenyl]-4-ylmethyl)-9-nitro-3,4,5,6-tetrahydro-2H-imidazo[2,1-b][1,3,6]oxadiazocine, was radiolabeled with the C-14 isotope. The synthesis of the (14)C-labeled nitroimidazole was accomplished in seven steps from diethanolamine with a final specific radioactivity of 3.552 GBq/mmol, a radiochemical yield of 87%, and a radiochemical purity of ≥96%. The source of the C-14 radiolabel was bromobenzene which was introduced by the Suzuki-Miyaura reaction. Tissue distribution results showed that the radiotracer has a high accumulation in the lungs of TB-infected mice, statistically significantly higher than in healthy mice. However, the clearance (for both TB-infected and non-TB-infected mice) from all organs (except the small intestine) from 1 to 2 h as well as the low percentage of injected dose per gram values achieved indicates breakdown of the compound in vivo and subsequent clearance from the body. The latter suggests that the compound might not be useful as an anti-TB drug in humans.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Carbenicillin disodium salt, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Trimethoprim lactate salt, ≥98%
Sigma-Aldrich
Carbenicillin disodium salt, 89.0-100.5% anhydrous basis
Sigma-Aldrich
Carbenicillin disodium, meets USP testing specifications
Carbenicillin sodium, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.