Merck
  • Rapamycin and everolimus facilitate hepatitis E virus replication: revealing a basal defense mechanism of PI3K-PKB-mTOR pathway.
Rapamycin and everolimus facilitate hepatitis E virus replication: revealing a basal defense mechanism of PI3K-PKB-mTOR pathway.
Journal of hepatology (2014-05-27)
Xinying Zhou, Yijin Wang, Herold J Metselaar, Harry L A Janssen, Maikel P Peppelenbosch, Qiuwei Pan
ABSTRACT
Humans are frequently exposed to hepatitis E virus (HEV). Nevertheless, the disease mainly affects pregnant women and immunocompromised individuals. Organ recipients receiving immunosuppressants, such as rapalogs, to prevent rejection have a high risk for developing chronic hepatitis following HEV infection. Rapalogs constitute potent inhibitors of mTOR including rapamycin and everolimus. As a master kinase, the mechanism-of-action of mTOR is not only associated with the immunosuppressive capacity of rapalogs but is also tightly regulated during pregnancy because of increased nutritional demands. We thus investigated the role of mTOR in HEV infection by using two state-of-the-art cell culture models: a subgenomic HEV containing luciferase reporter and a full-length HEV infectious cell culture system. In both subgenomic and full-length HEV models, HEV infection was aggressively escalated by treatment of rapamycin or everolimus. Inhibition of mTOR was confirmed by Western blot showing the inhibition of its downstream target, S6 phosphorylation. Consistently, stable silencing of mTOR by lentiviral RNAi resulted in a significant increase in intracellular HEV RNA, suggesting an antiviral function of mTOR in HEV infection. By targeting a series of other up- and downstream elements of mTOR signaling, we further revealed an effective basal defense mechanism of the PI3K-PKB-mTOR pathway against HEV, which is through the phosphorylated eIF4E-binding protein 1 (4E-BP1), however independent of autophagy formation. The discovery that PI3K-PKB-mTOR pathway limits HEV infection through 4E-BP1 and acts as a gate-keeper in human HEV target cells bears significant implications in managing immunosuppression in HEV-infected organ transplantation recipients.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)