Merck
  • Enantioselective nano liquid chromatographic separation of racemic pharmaceuticals: a facile one-pot in situ preparation of lipase-based polymer monoliths in capillary format.
Enantioselective nano liquid chromatographic separation of racemic pharmaceuticals: a facile one-pot in situ preparation of lipase-based polymer monoliths in capillary format.
Chirality (2014-03-08)
Marwa Ahmed, Ashraf Ghanem
ABSTRACT
New affinity monolithic capillary columns of 150 µm internal diameter were prepared in situ fused glass capillary via either immobilization or encapsulation of Candida antarctica lipase B (CALB) on or within polymer monoliths, respectively. The immobilized lipase-based monoliths were prepared via copolymerization of 19.1% monomers (8.9% MMA and 10.2% GMA), 19.8% EDMA, and 61.1% porogens (54.2% formamide and 6.9% 1-propanol) w/w or 20% GMA, 20% EDMA, and 60% porogens (51.6% cyclohexanol and 8.4% 1-dodecanol) in the presence of AIBN (1%) as a radical initiator. This was followed by pumping a solution of lipase through the capillaries and rinsing with potassium phosphate buffer. On the other hand, the encapsulated lipase-based monoliths were prepared via copolymerization of 20% monomers (GMA), 20% EDMA, and 60% porogens (48% 1-propanol, 6% 1,4-butanediol) or 16.4% monomers (16% BuMA, 0.4% SPMA), 23.6% EDMA, and 60% porogens (36% 1-propanol, 18% 1,4-butanediol along with 6% lipase aqueous solution in potassium phosphate buffer. The prepared capillary columns were investigated for the enantioselective nano liquid chromatographic separation of a set of different classes of racemic pharmaceuticals, namely, α- and β-blockers, antiinflammatory drugs, antifungal drugs, dopamine antagonists, norepinephrine-dopamine reuptake inhibitors, catecholamines, sedative hypnotics, diuretics, antihistaminics, anticancer drugs, and antiarrhythmic drugs. Run-to-run repeatability was quite satisfactory. The encapsulated lipase-based capillary monolith showed better enantioselective separations of most of the investigated compounds. Baseline separation was achieved for alprenolol, atenolol, bromoglutithimide, carbuterol, chloropheneramine, cizolertine carbinol, 4-hydroxy-3-methoxymandelic acid, desmethylcizolertine, nomifensine, normetanephrine, and sulconazole under reversed phase chromatographic conditions. A speculation about the understanding of the chiral recognition mechanism of lipase-based monoliths toward the investigated pharmaceuticals is discussed.
Sigma-Aldrich
Methanol, NMR reference standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Acetone, analytical standard
Supelco
1-Propanol, analytical standard
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%