Merck
  • Nuclear localization of Kaiso promotes the poorly differentiated phenotype and EMT in infiltrating ductal carcinomas.
Nuclear localization of Kaiso promotes the poorly differentiated phenotype and EMT in infiltrating ductal carcinomas.
Clinical & experimental metastasis (2014-02-27)
Jacqueline Jones, Honghe Wang, Balasubramanyam Karanam, Shaniece Theodore, Windy Dean-Colomb, Danny R Welch, William Grizzle, Clayton Yates
ABSTRACT
The expression and biological consequences of Kaiso, a novel bi-modal transcription factor, in infiltrating ductal carcinomas (IDCs) have not been widely investigated. In the present study, we determined Kaiso expression and subcellular localization in 146 normal tissues, 376 IDCs, and 85 lymph node metastases. In IDCs, there was higher Kaiso expression in both the cytoplasmic and nuclear compartments, which correlated with age <48 (cytoplasmic p < 0.0093; nuclear p < 0.0001) and moderate differentiation (cytoplasmic p < 0.0042; nuclear p < 0.0001), as determined by Chi square analysis. However, only nuclear Kaiso correlated with poor prognostic factors, i.e., race (African Americans) (p < 0.0001), poor differentiation (p < 0.0001), and metastases (p < 0.0001). Nuclear Kaiso was also associated with worse overall survival (p < 0.0019), with African American patients displaying worse survival rates relative to Caucasian patients (p < 0.029). MCF-7 (non-metastatic), MDA-MB-468 (few metastases), and MDA-MB-231 (highly metastatic) breast cancer cells demonstrated increasing Kaiso levels, with more nuclear localization in the highly metastatic cell line. Over-expression of Kaiso in MCF-7 cells increased cell migration and invasion, but treatment of MDA-MB-468 and MDA-MB-231 cells with si-Kaiso decreased cell migration and invasion and induced expression of E-cadherin RNA and protein. E-cadherin re-expression was associated with a reversal of mesenchymal associated cadherins, N-cadherin and cadherin 11, as well as decreased vitamin expression. Further, Kaiso directly bound to methylated sequences in the E-cadherin promoter, an effect prevented by 5-aza-2-deoxycytidine. Immunofluorescence co-staining of poorly differentiated IDCs demonstrated that nuclear Kaiso is associated with a loss of E-cadherin expression. These findings support a role for Kaiso in promoting aggressive breast tumors.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Zbtb33
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
MISSION® esiRNA, targeting human ZBTB33
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%