Merck
New amphiphilic N-phosphoryl oligopeptides designed for gene delivery.
International journal of pharmaceutics (2014-04-09)
Yunfei Sun, Long Chen, Fude Sun, Xibo Tian, Shi-Zhong Luo
ABSTRACT
Gene therapy is a potent tool for the treatment of cancer and other gene defect diseases, which involves using DNA that encodes a functional, therapeutic gene to replace a mutated gene. However, the DNA transfection efficiency is restricted by its negative charges and low susceptibility to endonucleases which prevent them penetrating tissue and cellular membranes. Both viral and non-viral vectors have been used for gene delivery, but the former are limited by their immunogenicity, while the latter are less efficient than their viral counterpart. Cationic amphiphilic lipopeptides whose structures can be easily modified and transformed have been used as non-viral vectors in gene delivery system due to their low cytotoxicity and high transfection efficiency. In this study, a series of cationic amphiphilic N-phosphoryl oligopeptides with varied lengths of hydrophobic tails and oligopeptide headgroups (C12-K6, C14-K6, C16-K6, Chol-K6 and C12-H6) were synthesized and used as gene delivery vectors. The affinities, abilities to condense pDNA and transfection efficiencies of the K6-lipopeptides were better than those of the H6-lipopeptides. In addition, the hydrophobic chains of the lipopeptides also affected their transfection efficiencies. The K6-lipopeptide with a hydrophobic chain of twelve carbons (C12-K6) showed the highest transfection efficiency in all these synthetic lipopeptides. At an optimal P/N ratio of 20, C12-K6 showed comparable pDNA transfection efficiency to PEI-25k, a well-defined gene delivery vector, but the cytotoxicity of C12-K6 was much lower. With acceptable gene transfection efficiency and low cytotoxicity, this cationic amphiphilic lipopeptide will have promising applications in gene therapy.
Sigma-Aldrich
Fmoc-His(Trt)-OH, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Cholesteryl chloroformate, 95%
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ≥99%
Supelco
1-Tetradecanol, Selectophore, ≥99.0%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Tetradecanol, 97%
Sigma-Aldrich
Thioanisole, ReagentPlus®, ≥99%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Supelco
1-Dodecanol, analytical standard
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Myristyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Fmoc-Lys(Boc)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
USP
Cetyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
1-Dodecanol, Selectophore, ≥98.0%
Supelco
Cetyl alcohol, analytical standard
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
1,2-Ethanedithiol, ≥98.0% (GC)
Sigma-Aldrich
1,2-Ethanedithiol, technical grade, ≥90%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Cetyl alcohol, European Pharmacopoeia (EP) Reference Standard