Merck
  • The role of nano-sized manganese oxides in the oxygen-evolution reactions by manganese complexes: towards a complete picture.
The role of nano-sized manganese oxides in the oxygen-evolution reactions by manganese complexes: towards a complete picture.
Dalton transactions (Cambridge, England : 2003) (2014-07-22)
Mohammad Mahdi Najafpour, Małgorzata Hołyńska, Amir Nasser Shamkhali, Sayed Habib Kazemi, Warwick Hillier, Emad Amini, M Ghaemmaghami, Davood Jafarian Sedigh, Atefeh Nemati Moghaddam, Rahim Mohamadi, Sasan Zaynalpoor, Katrin Beckmann
ABSTRACT
Eighteen Mn complexes with N-donor and carboxylate ligands have been synthesized and characterized. Three Mn complexes among them are new and are reported for the first time. The reactions of oxygen evolution in the presence of oxone (2KHSO5·KHSO4·K2SO4) and cerium(iv) ammonium nitrate catalyzed by these complexes are studied and characterized by UV-visible spectroscopy, X-ray diffraction spectrometry, dynamic light scattering, Fourier transform infrared spectroscopy, electron paramagnetic resonance spectroscopy, transmission electron microscopy, scanning electron microscopy, membrane-inlet mass spectrometry and electrochemistry. Some of these complexes evolve oxygen in the presence of oxone as a primary oxidant. CO2 and MnO4(-) are other products of these reactions. Based on spectroscopic studies, the true catalysts for oxygen evolution in these reactions are different. We proposed that for the oxygen evolution reactions in the presence of oxone, the true catalysts are both high valent Mn complexes and Mn oxides, but for the reactions in the presence of cerium(iv) ammonium nitrate, the active catalyst is most probably a Mn oxide.
Product Description
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Supelco
Di-n-octyl phthalate, analytical standard
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%