Merck
  • HRAS mutations and resistance to the epidermal growth factor receptor tyrosine kinase inhibitor erlotinib in head and neck squamous cell carcinoma cells.
HRAS mutations and resistance to the epidermal growth factor receptor tyrosine kinase inhibitor erlotinib in head and neck squamous cell carcinoma cells.
Head & neck (2013-10-15)
J Hun Hah, Mei Zhao, Curtis R Pickering, Mitchell J Frederick, Genevieve A Andrews, Samar A Jasser, David R Fooshee, Zvonimir L Milas, Chad Galer, Daisuke Sano, William N William, Edward Kim, John Heymach, Lauren A Byers, Vali Papadimitrakopoulou, Jeffrey N Myers
ABSTRACT
The purpose of this study was to identify mechanisms of innate resistance to an epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor, erlotinib, in a panel of head and neck squamous cell carcinoma (HNSCC) cell lines. Specifically, we analyzed the role of HRAS mutations in erlotinib resistance. Erlotinib sensitivity was determined by methyl thiazolyl-tetrazolium (MTT) assays. Molecular signaling pathways and somatic mutations were examined. Changes in sensitivity after modulation of HRAS expression were evaluated. All 7 cell lines were wild-type for EGFR and KRAS regardless of erlotinib sensitivity; however, 1 erlotinib-resistant cell line (HN31) harbored an HRAS G12D mutation. Downregulation of HRAS expression by small interfering RNA (siRNA) or short hairpin RNA (shRNA) in HN31 led to increased erlotinib sensitivity in vitro and in vivo. Transfection of activating HRAS-mutant (G12D and G12V) constructs into erlotinib-sensitive cell lines made them more resistant to erlotinib. Activating HRAS mutations can confer erlotinib resistance in an HRAS mutant HNSCC cell line.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hras1
Sigma-Aldrich
MISSION® esiRNA, targeting human HRAS
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%