Merck
RNA binding protein HuR regulates the expression of ABCA1.
Journal of lipid research (2014-04-15)
Cristina M Ramírez, Chin Sheng Lin, Kotb Abdelmohsen, Leigh Goedeke, Je-Hyun Yoon, Julio Madrigal-Matute, Jose L Martin-Ventura, Dat T Vo, Philip J Uren, Luiz O Penalva, Myriam Gorospe, Carlos Fernández-Hernando
ABSTRACT
ABCA1 is a major regulator of cellular cholesterol efflux and plasma HDL biogenesis. Even though the transcriptional activation of ABCA1 is well established, the posttranscriptional regulation of ABCA1 expression is poorly understood. Here, we investigate the potential contribution of the RNA binding protein (RBP) human antigen R (HuR) on the posttranscriptional regulation of ABCA1 expression. RNA immunoprecipitation assays demonstrate a direct interaction between HuR and ABCA1 mRNA. We found that HuR binds to the 3' untranslated region of ABCA1 and increases ABCA1 translation, while HuR silencing reduces ABCA1 expression and cholesterol efflux to ApoA1 in human hepatic (Huh-7) and monocytic (THP-1) cells. Interestingly, cellular cholesterol levels regulate the expression, intracellular localization, and interaction between HuR and ABCA1 mRNA. Finally, we found that HuR expression was significantly increased in macrophages from human atherosclerotic plaques, suggesting an important role for this RBP in controlling macrophage cholesterol metabolism in vivo. In summary, we have identified HuR as a novel posttranscriptional regulator of ABCA1 expression and cellular cholesterol homeostasis, thereby opening new avenues for increasing cholesterol efflux from atherosclerotic foam macrophages and raising circulat-ing HDL cholesterol levels.
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
T0901317, ≥98%
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Elavl1
Sigma-Aldrich
MISSION® esiRNA, targeting human ELAVL1
Supelco
Cycloheximide, PESTANAL®, analytical standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
SAFC
Glycine
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Supelco
Pefabloc® SC, analytical standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)