Merck
  • Downregulation of HTATIP2 expression is associated with promoter methylation and poor prognosis in glioma.
Downregulation of HTATIP2 expression is associated with promoter methylation and poor prognosis in glioma.
Experimental and molecular pathology (2015-01-27)
Xingyu Dong, Qingshan Deng, Xiaohu Nie, Maoying Zhang, Weiqiang Jia, Chen Chen, Chunwei Xu, Ruxiang Xu
ABSTRACT
Glioma is an aggressive tumor with poor prognosis. Identification of precise prognostic marker and effective therapeutic target is important in the treatment of glioma. HTATIP2 is a novel tumor suppressor gene, which is frequently silenced by epigenetic mechanisms in many caners. However, the expression of HTATIP2 and how it is regulated in glioma are unknown. Hence, we assessed whether loss of HTATIP2 expression occurs in glioma, and, if so, what is the mechanism of such loss. We found that HTATIP2 expression was absent or diminished in primary gliomas compared with normal brain tissue. In vitro experiments showed that HTATIP2 expression could be restored via 5-aza-2'deoxycytidine treatment in U87 and U251 cell lines. Methyl-specific PCR indicated that the two cell lines and 60% primary gliomas carried aberrant methylated HTATIP2 alleles while normal brain tissue did not. Pyrosequencing confirmed these results and showed a higher density of methylation in the minimal promoter element, which contains four Sp1 binding sites in primary gliomas, than in normal brain tissue. Finally, we found that the overall survival was significantly higher in patients with positive HTATIP2 expression than those with loss of HTATIP2 expression. Overexpression of HTATIP2 inhibited glioma proliferation and growth in vitro. Taken together, the present study showed that loss of HTATIP2 expression was a frequent event in glioma and is associated with poor prognosis. Promoter methylation may be an underlying mechanism.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
MISSION® esiRNA, targeting human HTATIP2
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard