Merck
  • Eutectic, monotectic and immiscibility systems of nimesulide with water-soluble carriers: phase equilibria, solid-state characterisation and in-vivo/pharmacodynamic evaluation.
Eutectic, monotectic and immiscibility systems of nimesulide with water-soluble carriers: phase equilibria, solid-state characterisation and in-vivo/pharmacodynamic evaluation.
The Journal of pharmacy and pharmacology (2014-06-20)
Hamdy Abdelkader, Ossama Y Abdallah, Hesham Salem, Adam W G Alani, Raid G Alany
ABSTRACT
The solid-state interactions of fused mixtures nimesulide (ND) with polyethylene glycol (PEG) 4000, urea or mannitol were studied through constructing thaw-melt phase equilibrium diagrams. The solid-state characteristics were investigated using differential scanning calorimetry (DSC) and X-ray diffraction (XRD). Various types of interactions were identified such as the formation of a eutectic system of ND-PEG 4000, monotectic system of ND-urea and complete solid immiscibility of ND with mannitol. The effects of carrier concentrations on the equilibrium solubility and in-vitro dissolution characteristics were studied. Linear increases (R(2)  > 0.99) in the aqueous solubility of ND in various concentrations of PEG 4000 and urea were obtained, whereas mannitol did not exhibit any effect on the solubility of ND. Similar trends were obtained with the dissolution efficiency of the fused mixtures of ND with PEG 4000 and urea compared with the corresponding physical mixtures and untreated drug. The analgesic effects of untreated ND and the selected formulations were investigated by evaluating the drug's ability to inhibit the acetic acid-induced writhing response. The analgesic effect of ND in a eutectic mixture with PEG 4000 and a monotectic mixture with urea was potentiated by 3.2 and 2.7-fold respectively compared with the untreated drug.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard