Merck
  • Modified-release calcifediol effectively controls secondary hyperparathyroidism associated with vitamin D insufficiency in chronic kidney disease.
Modified-release calcifediol effectively controls secondary hyperparathyroidism associated with vitamin D insufficiency in chronic kidney disease.
American journal of nephrology (2015-01-13)
Stuart M Sprague, Arnold L Silva, Fahd Al-Saghir, Radhika Damle, Samir P Tabash, Martin Petkovich, Eric J Messner, Jay A White, Joel Z Melnick, Charles W Bishop
ABSTRACT
Vitamin D insufficiency drives secondary hyperparathyroidism (SHPT) and is associated with increased cardiovascular mortality in patients with chronic kidney disease (CKD). SHPT is poorly addressed by current vitamin D repletion options. The present study evaluated a novel investigational vitamin D repletion therapy: a modified-release (MR) formulation of calcifediol designed to raise serum 25-hydroxyvitamin D in a gradual manner to minimize the induction of CYP24 and, thereby, improve the SHPT control. This randomized, double-blind, placebo-controlled trial evaluated MR calcifediol in CKD subjects (n = 78) with plasma intact parathyroid hormone (iPTH) >70 pg/ml and serum total 25-hydroxyvitamin D <30 ng/ml. Subjects received daily treatment for six weeks with oral MR calcifediol (30, 60 or 90 µg) or a placebo. More than 90% of subjects treated with MR calcifediol achieved serum 25-hydroxyvitamin D levels ≥30 ng/ml versus 3% of subjects treated with placebo (p < 0.0001). Mean plasma iPTH decreased from baseline (140.3 pg/ml) by 20.9 ± 6.2% (SE), 32.8 ± 5.7 and 39.3 ± 4.3% in the 30, 60 and 90 µg dose groups, respectively, and increased 17.2 ± 7.8% in the pooled placebo group (p < 0.005). No clinically significant safety concerns arose during MR calcifediol treatment. Oral MR calcifediol appears safe and highly effective in treating SHPT associated with vitamin D insufficiency in CKD.
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 50 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, 98% (CP)