Merck
  • Pathway as a pharmacological target for herbal medicines: an investigation from reduning injection.
Pathway as a pharmacological target for herbal medicines: an investigation from reduning injection.
PloS one (2015-04-02)
Jianling Liu, Ke Sun, Chunli Zheng, Xuetong Chen, Wenjuan Zhang, Zhengzhong Wang, Piar Ali Shar, Wei Xiao, Yonghua Wang
ABSTRACT
As a rich natural resource for drug discovery, Traditional Chinese Medicine (TCM) plays an important role in complementary and alternative medical systems. TCM shows a daunting complexity of compounds featuring multi-components and multi-targets to cure diseases, which thus always makes it extremely difficult to systematically explain the molecular mechanisms adequately using routine methods. In the present work, to reveal the systematic mechanism of herbal formulae, we developed a pathway-based strategy by combining the pathways integrating, target selection, reverse drug targeting and network analysis together, and then exemplified it by Reduning injection (RDN), a clinically widely used herbal medicine injection, in combating inflammation. The anti-inflammatory effects exerted by the major ingredients of RDN at signaling pathways level were systematically investigated. More importantly, our predicted results were also experimentally validated. Our strategy provides a deep understanding of the pharmacological functions of herbal formulae from molecular to systematic level, which may lead to more successful applications of systems pharmacology for drug discovery and development.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Chlorogenic acid, primary reference standard
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture