Merck
  • Canonical androstenedione reduction is the predominant source of signaling androgens in hormone-refractory prostate cancer.
Canonical androstenedione reduction is the predominant source of signaling androgens in hormone-refractory prostate cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-04-29)
Matthew Fankhauser, Yuen Tan, Geoff Macintyre, Izhak Haviv, Matthew K H Hong, Anne Nguyen, John S Pedersen, Anthony J Costello, Christopher M Hovens, Niall M Corcoran
ABSTRACT
It has been recognized for almost a decade that concentrations of signaling androgens sufficient to activate the androgen receptor are present in castration-resistant prostate cancer tissue. The source of these androgens is highly controversial, with three competing models proposed. We, therefore, wished to determine the androgenic potential of human benign and malignant (hormone-naïve and treated) prostate tissue when incubated with various precursors and examine concomitant changes in enzyme expression. Freshly harvested prostate tissue [benign, hormone-naïve, and hormone-refractory prostate cancer (HRPC)] was incubated in excess concentrations of cholesterol, progesterone, DHEA, androstenedione, or testosterone for 96 hours, and steroid concentrations in the conditioned media measured by gas chromatography-mass spectroscopy. Changes in the expression of androgen synthetic and/or degradative enzymes were determined by expression microarray and qPCR. Significant changes were confirmed in an independent dataset. Of the precursor molecules tested, only incubation with androstenedione gave rise to significant concentrations of signaling androgens. Although this was observed in all tissue types, it occurred to a significantly greater degree in hormone-refractory compared with hormone-naïve cancer. Consistent with this, gene set enrichment analysis of the expression microarray data revealed significant upregulation of 17HSD17B activity, with overexpression of the canonical enzyme AKR1C3 confirmed by qPCR in the same samples and in a publicly available expression dataset. Importantly, we found no evidence to support a significant contribution from either the "backdoor" or "5-α dione" pathway. Reduction of androstenedione to testosterone by the canonical HSD17B AKR1C3 is the predominant source of signaling androgens in HRPC.
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%