Merck
  • Antioxidant-capacity-based models for the prediction of acrylamide reduction by flavonoids.
Antioxidant-capacity-based models for the prediction of acrylamide reduction by flavonoids.
Food chemistry (2014-08-31)
Jun Cheng, Xinyu Chen, Sheng Zhao, Yu Zhang
ABSTRACT
The aim of this study was to investigate the applicability of artificial neural network (ANN) and multiple linear regression (MLR) models for the estimation of acrylamide reduction by flavonoids, using multiple antioxidant capacities of Maillard reaction products as variables via a microwave food processing workstation. The addition of selected flavonoids could effectively reduce acrylamide formation, which may be closely related to the number of phenolic hydroxyl groups of flavonoids (R: 0.735-0.951, P<0.001). The rate of inhibition of acrylamide formation correlated well with the change of trolox equivalent antioxidant capacity (ΔTEAC) measured by DPPH (R(2)=0.833), ABTS (R(2)=0.860) or FRAP (R(2)=0.824) assay. Both ANN and MLR models could effectively serve as predictive tools for estimating the reduction of acrylamide affected by flavonoids. The current predictive model study provides a low-cost and easy-to-use approach to the estimation of rates at which acrylamide is degraded, while avoiding tedious sample pretreatment procedures and advanced instrumental analysis.
Sigma-Aldrich
D-(+)-Glucose monohydrate, meets analytical specification of Ph. Eur., BP, Ph Franç., 97.5-102.0% anhydrous basis(HPLC)
Supelco
Methanol, analytical standard
Millipore
D-(+)-Glucose monohydrate, suitable for microbiology, ≥99.0%
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Apigenin, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
D-(+)-Glucose monohydrate, tested according to Ph. Eur.
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%