Merck
Fumonisins in corn (Zea mays L.) from Southern Brazil.
Food additives & contaminants. Part B, Surveillance (2014-06-11)
Vildes M Scussel, Geovana D Savi, Lea Luzia Freitas Costas, José Junior Mendonça Xavier, Daniel Manfio, Karoline O Bittencourt, Kin Aguiar, Stephanie M Stein
ABSTRACT
A total of 232 samples of corn commercialised in Santa Catarina state, Southern Brazil (temperate zone climate), were evaluated from 2007 to 2012 for fumonisins (FBs: FB1 and FB2). Before performing this study, a FBs method with liquid chromatography and fluorescence detection (ex. 335; em. 440 nm) was validated first. FBs were detected in 46.6% (108 samples), with values ranging from 66 to 7832 µg kg(-1) for FB1 and 110 to 1201 µg kg(-1) for FB2. The number of contaminated corn samples for FB1 and FB2 varied and often presented contamination of FB1 only. Per year of analysis, the numbers were: n = 22/8(FB1/FB2), 44/5(FB1/FB2), 25/12(FB1/FB2), 4(FB1), 6(FB1) and 7(FB1) in 2007, 2008, 2009, 2010, 2011 and 2012, respectively. The contamination percentage was 42.3/15.4, 59.5/6.8, 43.8/21.1, 36.4, 35.3 and 33.3%, respectively, during these years. Consumers can be exposed to these mycotoxins and their health can be at risk through the consumption of contaminated corn.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis